accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2387505-58-8

IUPAC Name: (2S)-4-amino-N-benzyl-2-[(2S)-2-[({1-[(2,6-dichlorophenyl)methyl]-3-[(pyrrolidin-1-yl)methyl]-1H-indol-6-yl}carbamoyl)amino]-3-(3,4-difluorophenyl)propanamido]butanamide dihydrochloride
Synonyms: (2S)-4-amino-N-benzyl-2-[(2S)-2-[({1-[(2,6-dichlorophenyl)methyl]-3-(pyrrolidin-1-ylmethyl)indol-6-yl}carbamoyl)amino]-3-(3,4-difluorophenyl)propanamido]butanamide dihydrochloride
Molecular Weight (g/mol): 863.65
Molecular Formula: C41H45Cl4F2N7O3
InChi Key: MGZSVSHDIOPSBO-ZEUUMAKDSA-N
SMILES: Cl.Cl.NCC[C@H](NC(=O)[C@H](CC1=CC(F)=C(F)C=C1)NC(=O)NC1=CC=C2C(CN3CCCC3)=CN(CC3=C(Cl)C=CC=C3Cl)C2=C1)C(=O)NCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results