Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RWJ 56110, Tocris Bioscience™
SDP

Catalog No. 26141 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-141 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 26-141 Supplier R&D Systems Supplier No. 2614/1

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective PAR1 antagonist

Selective protease-activated receptor-1 (PAR1) antagonist; displays no activity at PAR2, PAR3, or PAR4 subtypes. Blocks thrombin-induced platelet aggregation and activation of MAPK in HUVECs. Also inhibits angiogenesis in a chick embryo angiogenesis model in vivo.

Chemical Identifiers

CAS 2387505-58-8
Molecular Formula C41H45Cl4F2N7O3
Molecular Weight (g/mol) 863.65
InChI Key MGZSVSHDIOPSBO-ZEUUMAKDSA-N
Synonym ?s-n-1s-3-amino-1-phenylmethyl amino carbonyl propyl-?-1-2,6-dichlorophenyl methyl-3-1-pyrrolidinylmethyl-1h-indol-6-yl amino carbonyl amino-3,4-difluoro-benzenepropanamide
PubChem CID 90488773
IUPAC Name (2S)-4-amino-N-benzyl-2-[(2S)-2-[({1-[(2,6-dichlorophenyl)methyl]-3-[(pyrrolidin-1-yl)methyl]-1H-indol-6-yl}carbamoyl)amino]-3-(3,4-difluorophenyl)propanamido]butanamide dihydrochloride
SMILES Cl.Cl.NCC[C@H](NC(=O)[C@H](CC1=CC(F)=C(F)C=C1)NC(=O)NC1=CC=C2C(CN3CCCC3)=CN(CC3=C(Cl)C=CC=C3Cl)C2=C1)C(=O)NCC1=CC=CC=C1

Specifications

Quantity 1 mg
Formula Weight 863.65
Percent Purity >96%
Chemical Name or Material RWJ 56110
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.