CAS RN 2390-59-2
Ethyl Violet
CAS: 2390-59-2 Molecular Formula: C31H42ClN3 Molecular Weight (g/mol): 492.15 MDL Number: MFCD00011765 InChI Key: JVICFMRAVNKDOE-UHFFFAOYSA-M Synonym: C.I. 42600; Basic Violet 4 PubChem CID: 16955 IUPAC Name: [4-[bis[4-(diethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;chloride SMILES: [Cl-].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1)N(CC)CC
Ethyl Violet, 80%, Thermo Scientific™
CAS: 2390-59-2 Molecular Formula: C31H42ClN3 Molecular Weight (g/mol): 492.15 MDL Number: MFCD00011765 InChI Key: JVICFMRAVNKDOE-UHFFFAOYSA-M Synonym: Basic Violet 4,C.I. 42600 PubChem CID: 16955 IUPAC Name: [4-[bis[4-(diethylamino)phenyl]methylidene]cyclohexa-2,5-dien-1-ylidene]-diethylazanium;chloride SMILES: [Cl-].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1)N(CC)CC
Ethyl Violet, MP Biomedicals
CAS: 2390-59-2 Molecular Formula: C31H42ClN3 Molecular Weight (g/mol): 492.15 MDL Number: MFCD00011765 InChI Key: JVICFMRAVNKDOE-UHFFFAOYSA-M Synonym: Basic Violet 4,Ethyl purple 6B PubChem CID: 16955 IUPAC Name: tris[4-(diethylamino)phenyl]methylium chloride SMILES: [Cl-].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1)N(CC)CC
Ethyl Violet 98.0+%, TCI America™
CAS: 2390-59-2 Molecular Formula: C31H42ClN3 Molecular Weight (g/mol): 492.15 MDL Number: MFCD00011765 InChI Key: JVICFMRAVNKDOE-UHFFFAOYSA-M Synonym: Basic Violet 4 PubChem CID: 16955 IUPAC Name: tris[4-(diethylamino)phenyl]methylium chloride SMILES: [Cl-].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1)N(CC)CC
Ethyl Violet, TCI America™
CAS: 2390-59-2 Molecular Formula: C31H42ClN3 Molecular Weight (g/mol): 492.15 MDL Number: MFCD00011765 InChI Key: JVICFMRAVNKDOE-UHFFFAOYSA-M Synonym: ethyl violet,basic violet 4,c.i. basic violet 4,ethyl violet ax,ethyl violet gga,ethyl crystal violet,lowacryl violet 4,shikiso acid brilliant blue 6b,ethyl violet biological stain,unii-94uz9rd7tj PubChem CID: 16955 IUPAC Name: tris[4-(diethylamino)phenyl]methylium chloride SMILES: [Cl-].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1)N(CC)CC