Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl Violet, MP Biomedicals

Catalog No. ICN15801625 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15801625 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15801625 Supplier MP Biomedicals, Inc Supplier No. 0215801625
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical compound suitable for laboratory applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2390-59-2
Molecular Formula C31H42ClN3
Molecular Weight (g/mol) 492.15
MDL Number MFCD00011765
InChI Key JVICFMRAVNKDOE-UHFFFAOYSA-M
Synonym Basic Violet 4, Ethyl purple 6B
PubChem CID 16955
IUPAC Name tris[4-(diethylamino)phenyl]methylium chloride
SMILES [Cl-].CCN(CC)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC)C1=CC=C(C=C1)N(CC)CC

Specifications

Assay Percent Range ∽80%
Color Dark Green
Quantity 25 g
Formula Weight 492.1
Chemical Name or Material Ethyl Violet
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.