accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrCH3PCH3CH3CH3

CAS RN 23906-97-0

IUPAC Name: tetraoctylphosphanium bromide
Synonyms: tetraoctylphosphanium bromide
Molecular Weight (g/mol): 563.77
Molecular Formula: C32H68BrP
InChi Key: QVBRLOSUBRKEJW-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCC[P+](CCCCCCCC)(CCCCCCCC)CCCCCCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results