Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetra-n-octylphosphonium Bromide 98.0+%, TCI America™
SDP

Catalog No. T165025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T165025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T165025G Supplier TCI America Supplier No. T165025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23906-97-0
Molecular Formula C32H68BrP
Molecular Weight (g/mol) 563.77
MDL Number MFCD00210101
InChI Key QVBRLOSUBRKEJW-UHFFFAOYSA-M
PubChem CID 3015167
IUPAC Name tetraoctylphosphanium bromide
SMILES [Br-].CCCCCCCC[P+](CCCCCCCC)(CCCCCCCC)CCCCCCCC

Specifications

Melting Point 42°C
Color White
Quantity 25 g
Formula Weight 563.77
Percent Purity ≥98.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material Tetra-n-octylphosphonium Bromide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.