accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2391-30-2

IUPAC Name: sodium 8-amino-3-(4-methylphenyl)-2,4-dioxo-3-azatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-7-sulfonate
Synonyms: sodium 8-amino-3-(4-methylphenyl)-2,4-dioxo-3-azatricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene-7-sulfonate
Molecular Weight (g/mol): 404.37
Molecular Formula: C19H13N2NaO5S
InChi Key: HYLDLLCHFLSKAG-UHFFFAOYSA-M
SMILES: [Na+].CC1=CC=C(C=C1)N1C(=O)C2=CC(=C(N)C3=CC=CC(C1=O)=C23)S([O-])(=O)=O
Quantity 
  • (1)
  • (2)
  • (1)
Color 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)