Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brilliant Sulphaflavine, Thermo Scientific™

Catalog No. AC153221000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC153221000 100 g
AC153220250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC153221000 Supplier Thermo Scientific Chemicals Supplier No. 153221000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2391-30-2
Molecular Formula C19H13N2NaO5S
Molecular Weight (g/mol) 404.372
InChI Key HYLDLLCHFLSKAG-UHFFFAOYSA-M
Synonym 1H-Benz[de]isoquinoline-5-sulfonic acid, monosodium salt
PubChem CID 23678860
ChEBI CHEBI:87226
IUPAC Name sodium;6-amino-2-(4-methylphenyl)-1,3-dioxobenzo[de]isoquinoline-5-sulfonate
SMILES CC1=CC=C(C=C1)N2C(=O)C3=CC=CC4=C3C(=CC(=C4N)S(=O)(=O)[O-])C2=O.[Na+]

Specifications

Formula Weight 418.41
Quantity 100 g
Chemical Name or Material Brilliant Sulphaflavine
EINECSNumber 219-238-3
RTECSNumber DE4098000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.