accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CPCH3PCH3CH3

CAS RN 23936-60-9

IUPAC Name: [2-(dimethylphosphanyl)ethyl]dimethylphosphane
Synonyms: [2-(dimethylphosphanyl)ethyl]dimethylphosphane
Molecular Weight (g/mol): 150.14
Molecular Formula: C6H16P2
InChi Key: ZKWQSBFSGZJNFP-UHFFFAOYSA-N
SMILES: CP(C)CCP(C)C
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)