Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,2-Bis(Dimethylphosphino)Ethane

Catalog No. 2619395G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-193-95G 5 g
26-193-91G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 26-193-95G Supplier Sigma Aldrich Supplier No. 2619395G
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,2-Bis(dimethylphosphino)ethane (dmpe) is a polydentate phosphine ligand. Synthesis of 1,2-bis(dimethylphosphino)ethane has been reported. It reacts with ZrCl2 to afford ZrCl4(dmpe)2 complexes.1,2-Bis(dimethylphosphino)ethane (dmpe) may be employed as ligand in the preparation of cis-arylmethyl(dmpe) nickel(II) complexes.

Specifications

CAS 23936-60-9
Density 0.9 g/mL at 25°C
Boiling Point 180°C
Molecular Formula C6H16P2
Linear Formula (CH3)2PCH2CH2P(CH3)2
Refractive Index n20/D 1.507
MDL Number MFCD00008511
Quantity 5 g
Synonym DMPE; Dimethyl-2-(dimethylphosphino)ethylphosphine; Ethylenebis(dimethylphosphine)
Molecular Weight (g/mol) 150.14 g/mol
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.