accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOONHOHNO

CAS RN 23945-44-0

IUPAC Name: 2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carboxylic acid
Synonyms: steviolbioside isoorotic acid
Molecular Weight (g/mol): 156.1
Molecular Formula: C5H4N2O4
InChi Key: ZXYAAVBXHKCJJB-UHFFFAOYSA-N
SMILES: OC(=O)C1=CNC(=O)NC1=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results