CAS RN 2396-63-6
1,6-Heptadiyne, 98%
CAS: 2396-63-6 Molecular Formula: C7H8 Molecular Weight (g/mol): 92.15 MDL Number: MFCD00014925 InChI Key: RSPZSDWVQWRAEF-UHFFFAOYSA-N Synonym: 1,6-heptadiyne,1,6-heptadiyne 6ci, 8ci, 9ci,heptane-1,6-diyne,acmc-1cin6,1,6-heptadiyne, 97+%,wln: 1uu5uu1,4-01-00-01121 beilstein handbook reference PubChem CID: 337121 IUPAC Name: hepta-1,6-diyne SMILES: C#CCCCC#C
1,6-Heptadiyne, 97%
CAS: 2396-63-6 Molecular Formula: C7H8 Molecular Weight (g/mol): 92.141 MDL Number: MFCD00014925 InChI Key: RSPZSDWVQWRAEF-UHFFFAOYSA-N Synonym: 1,6-heptadiyne,1,6-heptadiyne 6ci, 8ci, 9ci,heptane-1,6-diyne,acmc-1cin6,1,6-heptadiyne, 97+%,wln: 1uu5uu1,4-01-00-01121 beilstein handbook reference PubChem CID: 337121 IUPAC Name: hepta-1,6-diyne SMILES: C#CCCCC#C
1,6-Heptadiyne 98.0+%, TCI America™
CAS: 2396-63-6 Molecular Formula: C7H8 Molecular Weight (g/mol): 92.141 MDL Number: MFCD00014925 InChI Key: RSPZSDWVQWRAEF-UHFFFAOYSA-N Synonym: 1,6-heptadiyne,1,6-heptadiyne 6ci, 8ci, 9ci,heptane-1,6-diyne,acmc-1cin6,1,6-heptadiyne, 97+%,wln: 1uu5uu1,4-01-00-01121 beilstein handbook reference PubChem CID: 337121 IUPAC Name: hepta-1,6-diyne SMILES: C#CCCCC#C
Sigma Aldrich 1,6-Heptadiyne
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
1,6-Heptadiyne, Thermo Scientific™
CAS: 2396-63-6 Molecular Formula: C7H8 Molecular Weight (g/mol): 92.141 MDL Number: MFCD00014925 InChI Key: RSPZSDWVQWRAEF-UHFFFAOYSA-N Synonym: 1,6-heptadiyne,1,6-heptadiyne 6ci, 8ci, 9ci,heptane-1,6-diyne,acmc-1cin6,1,6-heptadiyne, 97+%,wln: 1uu5uu1,4-01-00-01121 beilstein handbook reference PubChem CID: 337121 IUPAC Name: hepta-1,6-diyne SMILES: C#CCCCC#C