CAS RN 23964-57-0
IUPAC Name:
methyl 4-methyl-3-[2-(propylamino)propanamido]thiophene-2-carboxylate hydrochloride
Synonyms:
articaine hydrochloride
Molecular Weight (g/mol):
320.83
Molecular Formula:
C13H21ClN2O3S
InChi Key:
GDWDBGSWGNEMGJ-UHFFFAOYNA-N
SMILES:
Cl.CCCNC(C)C(=O)NC1=C(SC=C1C)C(=O)OC
1
–
2
of
2
results
2
Articaine Hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards