Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Articaine hydrochloride, 98%

Catalog No. AAH3685203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH36852-03 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH36852-03 Supplier Thermo Scientific™ Supplier No. H3685203
Add to Cart

Specifications

CAS 23964-57-0
Melting Point 173-180°
Molecular Formula C13H21ClN2O3S
MDL Number MFCD00190157
Quantity 1 g
InChI Key GDWDBGSWGNEMGJ-UHFFFAOYNA-N
SMILES Cl.CCCNC(C)C(=O)NC1=C(SC=C1C)C(=O)OC
IUPAC Name methyl 4-methyl-3-[2-(propylamino)propanamido]thiophene-2-carboxylate hydrochloride
Molecular Weight (g/mol) 320.83
Formula Weight 320.83
Percent Purity 98%
Physical Form Solid
Show More Show Less
ShelfLife 5 Years
EINECSNumber 245-957-7
RTECSNumber XM8511000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.