accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2399-48-6

IUPAC Name: (oxolan-2-yl)methyl prop-2-enoate
Synonyms: oxolan-2-ylmethyl prop-2-enoate
Molecular Weight (g/mol): 156.18
Molecular Formula: C8H12O3
InChi Key: YNXCGLKMOXLBOD-UHFFFAOYNA-N
SMILES: C=CC(=O)OCC1CCCO1
Boiling Point 
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)