Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetrahydrofurfuryl acrylate

Catalog No. 408271100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-827-1100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-827-1100ML Supplier Sigma Aldrich Supplier No. 408271100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetrahydrofurfuryl acrylate may be used as an acrylic matrix for silver nanoparticles/polymer nanocomposites . It can form copolymers with butadiene. Prior to transfer printing, cellulosic and proteinic fibers are grafted with THFA.

Specifications

CAS 2399-48-6
Density 1.064 g/mL (at 25°C)
Boiling Point 87°C (9 mmHg)
Molecular Formula C8H12O3
Refractive Index n20/D 1.46 (literature)
Linear Formula C8H12O3
Quantity 100 mL
MDL Number MFCD00014516
Synonym THFA
Molecular Weight (g/mol) 156.18
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.