accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 23996-16-9

H3CNNN

CAS RN 23996-16-9

IUPAC Name: 3-(2-undecyl-1H-imidazol-1-yl)propanenitrile
Synonyms: 3-(2-undecylimidazol-1-yl)propanenitrile
Molecular Weight (g/mol): 275.44
Molecular Formula: C17H29N3
InChi Key: SZUPZARBRLCVCB-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC1=NC=CN1CCC#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results