accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNN

CAS RN 23996-55-6

IUPAC Name: 3-(2-methyl-1H-imidazol-1-yl)propanenitrile
Synonyms: 3-(2-methylimidazol-1-yl)propanenitrile
Molecular Weight (g/mol): 135.17
Molecular Formula: C7H9N3
InChi Key: SESYNEDUKZDRJL-UHFFFAOYSA-N
SMILES: CC1=NC=CN1CCC#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results