Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Cyanoethyl)-2-methylimidazole 98.0+%, TCI America™
SDP

Catalog No. C196325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C196325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C196325G Supplier TCI America Supplier No. C196325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23996-55-6
Molecular Formula C7H9N3
Molecular Weight (g/mol) 135.17
InChI Key SESYNEDUKZDRJL-UHFFFAOYSA-N
Synonym 3-(2-Methyl-1-imidazolyl)propionitrile
PubChem CID 90328
IUPAC Name 3-(2-methylimidazol-1-yl)propanenitrile
SMILES CC1=NC=CN1CCC#N

Specifications

Melting Point 57°C
Color Yellow
pH ∼9.70 (0.5% H2O 25°C)
Quantity 25 g
UN Number 3439
Formula Weight 135.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(2-Cyanoethyl)-2-methylimidazole
EINECSNumber (5)-0398&(5)-0401
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.