accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2403-61-4

IUPAC Name: 1-chloro-4-(diethoxymethyl)benzene
Synonyms: 1-chloro-4-(diethoxymethyl)benzene
Molecular Weight (g/mol): 214.69
Molecular Formula: C11H15ClO2
InChi Key: GPLBEXMSGZWIPD-UHFFFAOYSA-N
SMILES: CCOC(OCC)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results