Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chlorobenzaldehyde Diethyl Acetal 97.0+%, TCI America™
SDP

Catalog No. C20445G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C20445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C20445G Supplier TCI America Supplier No. C20445G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2403-61-4
Molecular Formula C11H15ClO2
Molecular Weight (g/mol) 214.69
MDL Number MFCD08276331
InChI Key GPLBEXMSGZWIPD-UHFFFAOYSA-N
Synonym 1-Chloro-4-(diethoxymethyl)benzene
PubChem CID 326160
IUPAC Name 1-chloro-4-(diethoxymethyl)benzene
SMILES CCOC(OCC)C1=CC=C(Cl)C=C1

Specifications

Color Yellow
Boiling Point 129°C
Quantity 5 g
Formula Weight 214.69
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Chlorobenzaldehyde Diethyl Acetal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.