accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2407-68-3

IUPAC Name: (2E)-3-(dimethylamino)prop-2-enenitrile
Synonyms: 3-dimethylaminoacrylonitrile
Molecular Weight (g/mol): 96.13
Molecular Formula: C5H8N2
InChi Key: ZKKBIZXAEDFPNL-HWKANZROSA-N
SMILES: CN(C)\C=C\C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results