Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Dimethylamino)acrylonitrile 97.0+%, TCI America™
SDP

Catalog No. D209425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D209425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D209425G Supplier TCI America Supplier No. D209425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2407-68-3
Molecular Formula C5H8N2
Molecular Weight (g/mol) 96.133
MDL Number MFCD00001929
InChI Key ZKKBIZXAEDFPNL-HYXAFXHYSA-N
PubChem CID 6436777
IUPAC Name (Z)-3-(dimethylamino)prop-2-enenitrile
SMILES CN(C)C=CC#N

Specifications

Boiling Point 115°C
Quantity 25 g
UN Number 2810
Formula Weight 96.13
Percent Purity ≥97.0% (N)
Physical Form Liquid
Chemical Name or Material 3-(Dimethylamino)acrylonitrile
RTECSNumber UD1302500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.