accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24070-77-7

IUPAC Name: 2-methylcyclopentan-1-ol
Synonyms: cyclopentanol, 2-methyl-, cis-
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: BVIJQMCYYASIFP-UHFFFAOYNA-N
SMILES: CC1CCCC1O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results