Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Methylcyclopentanol-1, MP Biomedicals

Catalog No. ICN225160 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN225160 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN225160 Supplier MP Biomedicals, Inc Supplier No. 0522516005

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 24070-77-7
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00066162
InChI Key BVIJQMCYYASIFP-UHFFFAOYNA-N
Synonym 2-methylcyclopentanol, 2-methyl cyclopentanol, cyclopentanol, 2-methyl, 2-methylcyclopentyl alcohol, cyclopentanol, 2-methyl-, cis, 2-methycyclopentanol, acmc-20ajgt, acmc-1ccvg, 2-methyl-1-cyclopentanol, 2-methylcyclopentanol-1
PubChem CID 32205
IUPAC Name 2-methylcyclopentan-1-ol
SMILES CC1CCCC1O

Specifications

Quantity 5 g
Formula Weight 100.16
Chemical Name or Material 2-Methylcyclopentanol-1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.