accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24093-81-0

IUPAC Name: methyl 3,5-dihydroxy-4-methoxybenzoate
Synonyms: methyl 3,5-dihydroxy-4-methoxybenzoate
Molecular Weight (g/mol): 198.17
Molecular Formula: C9H10O5
InChi Key: SUGIJIFASYORQN-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(O)=C(OC)C(O)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results