Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3,5-Dihydroxy-4-methoxybenzoate 98.0+%, TCI America™
SDP

Catalog No. M27945G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27945G 5 g
M27941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27945G Supplier TCI America Supplier No. M27945G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 24093-81-0
Molecular Formula C9H10O5
Molecular Weight (g/mol) 198.17
MDL Number MFCD00017195
InChI Key SUGIJIFASYORQN-UHFFFAOYSA-N
Synonym 3,5-Dihydroxy-4-methoxybenzoic Acid Methyl Ester
PubChem CID 5319726
IUPAC Name methyl 3,5-dihydroxy-4-methoxybenzoate
SMILES COC(=O)C1=CC(O)=C(OC)C(O)=C1

Specifications

Melting Point 149°C
Color White
Quantity 5 g
Formula Weight 198.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 3,5-Dihydroxy-4-methoxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.