accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONBr

CAS RN 24100-18-3

IUPAC Name: 2-bromo-3-methoxypyridine
Synonyms: 2-bromo-3-methoxypyridine
Molecular Weight (g/mol): 188.02
Molecular Formula: C6H6BrNO
InChi Key: PDOWLYNSFYZIQX-UHFFFAOYSA-N
SMILES: COC1=CC=CN=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results