accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2411-83-8

IUPAC Name: methyl 2,3-dihydroxybenzoate
Synonyms: methyl 2,3-dihydroxybenzoate
Molecular Weight (g/mol): 168.15
Molecular Formula: C8H8O4
InChi Key: DOAJWTSNTNAEIY-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(O)=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results