Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,3-Dihydroxybenzoate 98.0+%, TCI America™
SDP

Catalog No. M17375G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17375G 5 g
M173725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17375G Supplier TCI America Supplier No. M17375G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2411-83-8
Molecular Formula C8H8O4
Molecular Weight (g/mol) 168.148
InChI Key DOAJWTSNTNAEIY-UHFFFAOYSA-N
Synonym o-pyrocatechuic acid, methyl ester, methyl2,3-dihydroxybenzoate, 2,3-dihydroxybenzoic acid methyl ester, benzoic acid, 2,3-dihydroxy-, methyl ester, 2,3-dihydroxy-benzoic acid methyl ester, acmc-1ckr1, methyl 2,3-dihydroxy-benzoate, rarechem al bf 0035, 2,3-dihydroxybenzoic acid methyl, dihydroxy benzoic acid methyl ester
PubChem CID 75484
IUPAC Name methyl 2,3-dihydroxybenzoate
SMILES COC(=O)C1=C(C(=CC=C1)O)O

Specifications

Melting Point 81°C
Boiling Point 96°C
Quantity 5 g
Formula Weight 168.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2,3-Dihydroxybenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.