accessibility menu, dialog, popup

UserName

Viewing profile as user:
SPSS

CAS RN 24171-89-9

IUPAC Name: tris(thiophen-2-yl)phosphane
Synonyms: tris(thiophen-2-yl)phosphane
Molecular Weight (g/mol): 280.36
Molecular Formula: C12H9PS3
InChi Key: KUCPTMZJPDVWJL-UHFFFAOYSA-N
SMILES: S1C=CC=C1P(C1=CC=CS1)C1=CC=CS1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)