Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tris(2-thienyl)phosphine, 98+%

Catalog No. AAA1337906
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1337906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1337906 Supplier Thermo Scientific Chemicals Supplier No. A1337906

missing translation for 'validMainframeMsg'

CAS: 24171-89-9 | C12H9PS3 | 280.36 g/mol

It employed as a intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It employed as a intermediate for pharmaceutical and organic synthesis.

Solubility
Insoluble in water.

Notes
Air Sensitive, store away from air. Store in cool, dry place in tightly closed container. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24171-89-9
Molecular Formula C12H9PS3
Molecular Weight (g/mol) 280.36
MDL Number MFCD00015471
InChI Key KUCPTMZJPDVWJL-UHFFFAOYSA-N
Synonym tris 2-thienyl phosphine, phosphine, tri-2-thienyl, tri 2-thienyl phosphine, tri thiophen-2-yl phosphine, tri2-thienylphosphine, tris thiophen-2-yl phosphane, tri-2-thienylphosphine, trithienylphosphine, pubchem6427, tri 2-thienyl phosphine #
PubChem CID 90384
IUPAC Name trithiophen-2-ylphosphane
SMILES S1C=CC=C1P(C1=CC=CS1)C1=CC=CS1

Specifications

Melting Point 28°C to 30°C
Odor Recognizable
Quantity 5 g
Beilstein 203736
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 280.37
Percent Purity ≥98%
Chemical Name or Material Tris(2-thienyl)phosphine
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 246-059-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.