accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2418-95-3

IUPAC Name: (2S)-2-azaniumyl-6-{[(tert-butoxy)carbonyl]amino}hexanoate
Synonyms: (2S)-2-ammonio-6-[(tert-butoxycarbonyl)amino]hexanoate
Molecular Weight (g/mol): 246.31
Molecular Formula: C11H22N2O4
InChi Key: VVQIIIAZJXTLRE-QMMMGPOBSA-N
SMILES: CC(C)(C)OC(=O)NCCCC[C@H]([NH3+])C([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (3)
Percent Purity 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)