Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nepsilon-(tert-Butoxycarbonyl)-L-lysine 97.0+%, TCI America™
SDP

Catalog No. B16705G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16705G 5 g
B16701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16705G Supplier TCI America Supplier No. B16705G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2418-95-3 | C11H22N2O4 | 246.31 g/mol

Chemical Identifiers

CAS 2418-95-3
Molecular Formula C11H22N2O4
Molecular Weight (g/mol) 246.31
MDL Number MFCD00037221
InChI Key VVQIIIAZJXTLRE-QMMMGPOBSA-N
Synonym h-lys boc-oh, lys boc, nepsilon-tert-butoxycarbonyl-l-lysine, n6-boc-l-lysine, n∼6∼-tert-butoxycarbonyl-l-lysine, nepsilon-boc-l-lysine, ne-boc-l-lysine, n epsilon-boc-l-lysine, h-lys boc-2-chlorotrityl resin, n6-tert-butoxycarbonyl-l-lysine
PubChem CID 2733283
IUPAC Name (2S)-2-azaniumyl-6-{[(tert-butoxy)carbonyl]amino}hexanoate
SMILES CC(C)(C)OC(=O)NCCCC[C@H]([NH3+])C([O-])=O

Specifications

Color White
Quantity 5 g
Formula Weight 246.31
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Nepsilon-(tert-Butoxycarbonyl)-L-lysine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.