accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3ON

CAS RN 24197-95-3

IUPAC Name: 4-[2-(dimethylamino)ethoxy]benzonitrile
Synonyms: 4-[2-(dimethylamino)ethoxy]benzonitrile
Molecular Weight (g/mol): 190.25
Molecular Formula: C11H14N2O
InChi Key: DTYDLVIYHGGCOG-UHFFFAOYSA-N
SMILES: CN(C)CCOC1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results