Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-[2-(Dimethylamino)ethoxy]benzonitrile, ≥97%, Thermo Scientific™

Catalog No. CC51516DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC51516DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC51516DA Supplier Thermo Scientific Chemicals Supplier No. CC51516DA
Add to Cart

Chemical Identifiers

CAS 24197-95-3
Molecular Formula C11H14N2O
Molecular Weight (g/mol) 190.25
MDL Number MFCD05738937
InChI Key DTYDLVIYHGGCOG-UHFFFAOYSA-N
Synonym 4-2-dimethylamino ethoxy benzonitrile, benzonitrile, 4-2-dimethylamino ethoxy, 4-2-dimethylaminoethoxy benzonitrile, 4-2-dimethylaminoethyloxy benzonitrile, 4-2-n,n-dimethylamino ethoxy benzonitrile, 4-2-dimethylamino ethoxy benzenecarbonitrile
PubChem CID 546912
IUPAC Name 4-[2-(dimethylamino)ethoxy]benzonitrile
SMILES CN(C)CCOC1=CC=C(C=C1)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 190.24
Percent Purity 97%
Chemical Name or Material 4-[2-(Dimethylamino)ethoxy]benzonitrile
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Causes serious eye damage.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.