accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CONH3CONCH3ONHCH3NH2CH3H3CONH3CONCH3ONHCH3NH2OOHOOH(E/Z)(E/Z)(E/Z)

CAS RN 242143-80-2

IUPAC Name: bis(5-amino-N,5-dimethyl-N-(1-{methyl[1-(methylcarbamoyl)-2-phenylethyl]carbamoyl}-2-(naphthalen-2-yl)ethyl)hex-2-enamide); but-2-enedioic acid
Synonyms: bis(5-amino-N,5-dimethyl-N-(1-{methyl[1-(methylcarbamoyl)-2-phenylethyl]carbamoyl}-2-(naphthalen-2-yl)ethyl)hex-2-enamide); butenedioic acid
Molecular Weight (g/mol): 1173.47
Molecular Formula: C68H84N8O10
InChi Key: YQDJPWXYLLPOPW-UHFFFAOYNA-N
SMILES: OC(=O)C=CC(O)=O.CNC(=O)C(CC1=CC=CC=C1)N(C)C(=O)C(CC1=CC=C2C=CC=CC2=C1)N(C)C(=O)C=CCC(C)(C)N.CNC(=O)C(CC1=CC=CC=C1)N(C)C(=O)C(CC1=CC=C2C=CC=CC2=C1)N(C)C(=O)C=CCC(C)(C)N
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)