accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOOHOOONN(S)

CAS RN 242478-38-2

IUPAC Name: 1-azabicyclo[2.2.2]octan-3-yl (1S)-1-phenyl-1,2,3,4-tetrahydroisoquinoline-2-carboxylate; butanedioic acid
Synonyms: 1-azabicyclo[2.2.2]octan-3-yl (1S)-1-phenyl-3,4-dihydro-1H-isoquinoline-2-carboxylate; succinic acid
Molecular Weight (g/mol): 480.56
Molecular Formula: C27H32N2O6
InChi Key: RXZMMZZRUPYENV-DZSUWJOWSA-N
SMILES: OC(=O)CCC(O)=O.O=C(OC1CN2CCC1CC2)N1CCC2=CC=CC=C2[C@@H]1C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results