Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Solifenacin Succinate 98.0+%, TCI America™
SDP

Catalog No. S094425MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S094425MG 25 mg
S0944100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S094425MG Supplier TCI America Supplier No. S094425MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 242478-38-2
Molecular Formula C27H32N2O6
Molecular Weight (g/mol) 480.56
MDL Number MFCD09952287
InChI Key RXZMMZZRUPYENV-DZSUWJOWSA-N
PubChem CID 133687147
IUPAC Name 1-azabicyclo[2.2.2]octan-3-yl (1S)-1-phenyl-1,2,3,4-tetrahydroisoquinoline-2-carboxylate; butanedioic acid
SMILES OC(=O)CCC(O)=O.O=C(OC1CN2CCC1CC2)N1CCC2=CC=CC=C2[C@@H]1C1=CC=CC=C1

Specifications

Melting Point 147°C
Color White
Quantity 25 mg
Formula Weight 480.56
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Solifenacin Succinate
RTECSNumber NW7136000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.