accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2425-74-3

IUPAC Name: N-tert-butylformamide
Synonyms: N-tert-butylformamide
Molecular Weight (g/mol): 101.15
Molecular Formula: C5H11NO
InChi Key: SDLAKRCBYGZJRW-UHFFFAOYSA-N
SMILES: CC(C)(C)NC=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results