Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-tert-Butylformamide 98.0+%, TCI America™
SDP

Catalog No. B127325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B127325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B127325ML Supplier TCI America Supplier No. B127325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2425-74-3
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00003277
InChI Key SDLAKRCBYGZJRW-UHFFFAOYSA-N
Synonym N-Formyl-tert-butylamine
PubChem CID 17045
IUPAC Name N-tert-butylformamide
SMILES CC(C)(C)NC=O

Specifications

Color Yellow
Boiling Point 202°C
Quantity 25 mL
Formula Weight 101.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-tert-Butylformamide
RTECSNumber LQ1450000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.