accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2425-79-8

IUPAC Name: 2-({4-[(oxiran-2-yl)methoxy]butoxy}methyl)oxirane
Synonyms: 2-{[4-(oxiran-2-ylmethoxy)butoxy]methyl}oxirane
Molecular Weight (g/mol): 202.25
Molecular Formula: C10H18O4
InChi Key: SHKUUQIDMUMQQK-UHFFFAOYNA-N
SMILES: C(CCOCC1CO1)COCC1CO1
Boiling Point 
  • (1)
  • (2)
  • (5)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (5)
  • (2)
  • (3)
  • (1)
  • (2)
  • (5)
  • (2)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)