Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Butanediol Diglycidyl Ether 93.0+%, TCI America™
SDP

Catalog No. B096425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B096425ML 25 mL
B0964250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B096425ML Supplier TCI America Supplier No. B096425ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2425-79-8
Molecular Formula C10H18O4
Molecular Weight (g/mol) 202.25
MDL Number MFCD00005146
InChI Key SHKUUQIDMUMQQK-UHFFFAOYNA-N
Synonym 1,4-butanediol diglycidyl ether, 1,4-diglycidyloxybutane, 1,4-bis glycidyloxy butane, butanediol diglycidyl ether, araldit dy 026, 1,4-butane diglycidyl ether, grilonit rv 1806, 1,4-diglycidloxybutane, chs-rr2, 1,4-bis 2,3-epoxypropoxy butane
PubChem CID 17046
IUPAC Name 2-({4-[(oxiran-2-yl)methoxy]butoxy}methyl)oxirane
SMILES C(CCOCC1CO1)COCC1CO1

Specifications

Melting Point -21°C
Color Yellow
Boiling Point 160°C
Quantity 25 mL
Formula Weight 202.25
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Butanediol Diglycidyl Ether
EINECSNumber (2)-0396
RTECSNumber EJ5100000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.