accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24255-27-4

IUPAC Name: 2-(morpholin-4-yl)-3-nitropyridin-1-ium
Synonyms: 2-(morpholin-4-yl)-3-nitropyridin-1-ium
Molecular Weight (g/mol): 210.21
Molecular Formula: C9H12N3O3
InChi Key: QMINJWUSSJFQBR-UHFFFAOYSA-O
SMILES: [O-][N+](=O)C1=CC=C[NH+]=C1N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results