Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(3-Nitro-2-pyridinyl)morpholine, ≥97%, Thermo Scientific™

Catalog No. CC24625EA
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC24625EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC24625EA Supplier Thermo Scientific Chemicals Supplier No. CC24625EA

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 24255-27-4
Molecular Formula C9H12N3O3
Molecular Weight (g/mol) 210.21
MDL Number MFCD00452814
InChI Key QMINJWUSSJFQBR-UHFFFAOYSA-O
Synonym 4-3-nitropyridin-2-yl morpholine, 4-3-nitro-2-pyridinyl morpholine, 4-3-nitro-pyridin-2-yl-morpholine, 4-3-nitro-2-pyridyl morpholine, nitropyridinylmorpholine, cbmicro_018334, cambridge id 5341296, 4-3-nitropyridine-2-yl morpholine, morpholine,4-3-nitro-2-pyridinyl
PubChem CID 2795439
IUPAC Name 2-(morpholin-4-yl)-3-nitropyridin-1-ium
SMILES [O-][N+](=O)C1=CC=C[NH+]=C1N1CCOCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 209.2
Chemical Name or Material 4-(3-Nitro-2-pyridinyl)morpholine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.