accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24259-59-4

IUPAC Name: (3S,4R,5S)-5-(hydroxymethyl)oxolane-2,3,4-triol
Synonyms: L-ribofuranose
Molecular Weight (g/mol): 150.13
Molecular Formula: C5H10O5
InChi Key: HMFHBZSHGGEWLO-ZHZWXSSZNA-N
SMILES: OC[C@@H]1OC(O)[C@@H](O)[C@H]1O
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (3)
  • (2)
  • (2)
Physical Form 
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (4)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)