CAS RN 24269-96-3
IUPAC Name:
methyl 5-[2-(5-nitrofuran-2-yl)ethenyl]furan-2-carboxylate
Synonyms:
methyl 5-[2-(5-nitrofuran-2-yl)ethenyl]furan-2-carboxylate
Molecular Weight (g/mol):
263.21
Molecular Formula:
C12H9NO6
InChi Key:
YDJPHSNZGRVPCK-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=C(O1)C=CC1=CC=C(O1)[N+]([O-])=O
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ βARK1 Inhibitor
The βARK1 Inhibitor, also referenced under CAS 24269-96-3, controls the biological activity of βARK1. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
2
Thermo Scientific Chemicals beta-Adrenergic Receptor Kinase1 Inhibitor
CAS: 24269-96-3 Molecular Formula: C12H9NO6 Synonym: β-ARK1 Inhibitor; GRK2 Inhibitor