CAS RN 24271-12-3
IUPAC Name:
octadecyl docosanoate
Synonyms:
stearyl behenate
Molecular Weight (g/mol):
593.08
Molecular Formula:
C40H80O2
InChi Key:
GAQPWOABOQGPKA-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCCCCCCCCCCCC(=O)OCCCCCCCCCCCCCCCCCC
1
–
1
of
1
results
1
MP Biomedicals, Inc Stearyl Behenate, MP Biomedicals
CAS: 24271-12-3 Molecular Formula: C40H80O2 Molecular Weight (g/mol): 593.08 MDL Number: MFCD00048672 InChI Key: GAQPWOABOQGPKA-UHFFFAOYSA-N Synonym: stearyl behenate,docosanoic acid, octadecyl ester,unii-a68s9ui39d,octadecanyl docosanoate,behenic acid stearyl ester,docosanoic acid octadecyl ester,unii-7ari9lth0u component PubChem CID: 90435 IUPAC Name: octadecyl docosanoate SMILES: CCCCCCCCCCCCCCCCCCCCCC(=O)OCCCCCCCCCCCCCCCCCC