Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stearyl Behenate, MP Biomedicals

Catalog No. ICN218614 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN218614 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN218614 Supplier MP Biomedicals, Inc Supplier No. 0521861401

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 24271-12-3
Molecular Formula C40H80O2
Molecular Weight (g/mol) 593.08
MDL Number MFCD00048672
InChI Key GAQPWOABOQGPKA-UHFFFAOYSA-N
Synonym stearyl behenate, docosanoic acid, octadecyl ester, unii-a68s9ui39d, octadecanyl docosanoate, behenic acid stearyl ester, docosanoic acid octadecyl ester, unii-7ari9lth0u component
PubChem CID 90435
IUPAC Name octadecyl docosanoate
SMILES CCCCCCCCCCCCCCCCCCCCCC(=O)OCCCCCCCCCCCCCCCCCC

Specifications

Quantity 1 g
Chemical Name or Material Stearyl Behenate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.