accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 24292-52-2

H3COOOHO(S)O(R)O(R)O(S)CH3(R)HO(R)HO(R)OH(S)HO(S)OH(R)OHOCH3HO(E)

CAS RN 24292-52-2

IUPAC Name: (2E)-3-(3-hydroxy-4-methoxyphenyl)-1-(2-hydroxy-6-methoxy-4-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-({[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy}methyl)oxan-2-yl]oxy}phenyl)prop-2-en-1-one
Synonyms: hesperidin methyl chalcone
Molecular Weight (g/mol): 624.59
Molecular Formula: C29H36O15
InChi Key: FDHNLHLOJLLXDH-JIYHLSBYSA-N
SMILES: COC1=CC=C(\C=C\C(=O)C2=C(O)C=C(O[C@@H]3O[C@H](CO[C@@H]4O[C@@H](C)[C@H](O)[C@@H](O)[C@H]4O)[C@@H](O)[C@H](O)[C@H]3O)C=C2OC)C=C1O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results